Learn More
American Radiolabeled Chemicals Inc TELMISARTAN-3H(G) 50UC
SDP
Catalog No. 5090108493
Encompass_Preferred
Telmisartan-3H(G) 50uCi in ethanol, 1 mCi/ml, S.A. 5-15 Ci/mmol, M.W. 514.62, Exclusive
Catalog No. 50-901-08493 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 1484 50UC
Add to Cart
Add to Cart
Request a Quote
Telmisartan-3H(G) 50uCi in ethanol, 1 mCi/ml, S.A. 5-15 Ci/mmol, M.W. 514.62, Exclusive