Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Telmisartan, Tocris Bioscience™

Catalog No. 513910R
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-391-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-391-0R Supplier R&D Systems Supplier No. 5139/10
This item is not returnable. View return policy

AT1 antagonist; PPARγ partial agonist

Telmisartan is an antagonist of the angiotensin II type 1 receptor (AT1) (Ki = 3.7 nM). Exhibits antihypertensive activity in a renin-dependent model of hypertension. Partial agonist of PPARγ receptors; shown to improve insulin sensitivity in rodents receiving high fat diets.

Chemical Identifiers

CAS 144701-48-4
Molecular Formula C33H30N4O2
Molecular Weight (g/mol) 514.629
InChI Key RMMXLENWKUUMAY-UHFFFAOYSA-N
Synonym telmisartan, micardis, pritor, bibr 277, kinzalmono, bibr 277se, 4'-1,7'-dimethyl-2'-propyl-1h,3'h-2,5'-bibenzo d imidazol-3'-yl methyl-1,1'-biphenyl-2-carboxylic acid, bibr-277, targit, telmisartan inn
PubChem CID 65999
ChEBI CHEBI:9434
IUPAC Name 2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoic acid
SMILES CCCC1=NC2=C(C=C(C=C2N1CC3=CC=C(C=C3)C4=CC=CC=C4C(=O)O)C5=NC6=CC=CC=C6N5C)C

Specifications

Color White
Quantity 10 mg
Formula Weight 514.62
Percent Purity >99%
Physical Form Solid
Chemical Name or Material Telmisartan
Recommended Storage Store at RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.