Learn More
Cayman Chemical Telmisartan-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1189889-44-8, 1 mg
SDP
Catalog No. 502964078
Encompass_Preferred
An internal standard for the quantification of telmisartan by GC- or LC-MS
Catalog No. 50-296-4078 Supplier Cayman Chemical Supplier No. 225681 MG
An internal standard for the quantification of telmisartan by GC- or LC-MS