Learn More
Cayman Chemical TelmIsartn-d3 500ug
SDP
Catalog No. 502986922
Encompass_Preferred
An internal standard for the quantification of telmisartan by GC- or LC-MS
Catalog No. 50-298-6922 Supplier Cayman Chemical Supplier No. 22568500 UG
An internal standard for the quantification of telmisartan by GC- or LC-MS