Learn More
Cayman Chemical Telmisartan, Purity: ≥98%, CAS: 144701-48-4, 50 mg
SDP
Catalog No. 502954065
Encompass_Preferred
A nonpeptide angiotensin II receptor antagonist which selectively and insurmountably inhibits the angiotensin II AT1 receptor subtype (Ki = 3.7 nM); a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 μM); potently reduces blood pressure
Catalog No. 50-295-4065 Supplier Cayman Chemical Supplier No. 1161550 MG
A nonpeptide angiotensin II receptor antagonist which selectively and insurmountably inhibits the angiotensin II AT1 receptor subtype (Ki = 3.7 nM); a partial agonist of PPARγ, activating the receptor to 25-30% of that produced by the full agonist rosiglitazone (EC50 = 4.5 μM); potently reduces blood pressure