Learn More
Kactus TelN Protelomerase (TLN-BE001) 10KU
Catalog No. NC2681695
Encompass
TelN Protelomerase is cloned from bacteriophage N15. TelN cuts dsDNA at the recognition site TelRL (56 bp), which consists of a palindromic sequence-TelO in the middle and palindromic sequences R3 and L3 at both ends of 14 bp form. TelN has cutting-ligating enzyme activity and covalently ligates at the cleavage site forming hairpin termini.
Catalog No. NC2681695 Supplier Kactus Supplier No. TLNBE00110KU
TelN Protelomerase is cloned from bacteriophage N15. TelN cuts dsDNA at the recognition site TelRL (56 bp), which consists of a palindromic sequence-TelO in the middle and palindromic sequences R3 and L3 at both ends of 14 bp form. TelN has cutting-ligating enzyme activity and covalently ligates at the cleavage site forming hairpin termini.