Learn More
Advanced Biomatrix TeloCol-6, Solution, Acid Soluble, 6 mg/ml with Neutralization Solution, Telocollagen
Catalog No. 501960298
Encompass_Preferred
TeloCol-6 bovine collagen solution is derived from an acid extraction process yielding a telopeptide-intact collagen. The pro-peptide regions at both ends of the collagen chain, N- and C-telopeptide regions, are maintained. TeloCol-6 collagen is approximately 95% Type I collagen with the remainder being comprised of Type III collagen. Each product includes a bottle containing 50 ml of collagen solution accompanied with a bottle of pre-formulated neutralizing solution for the formation of a collagen gel. This product is supplied at a concentration of approximately 6 mg/ml with the concentration for each specific lot provided on the product label and Certificate of Analysis that is available with the purchase of each product. TeloCol-6 is sterile filtered and provided in user-friendly packaging for use and storage. TeloCol-6 is ideal for coating of surfaces, providing preparation of thin layers for culturing cells, or use as a solid gel. Collagen Video: https://youtu.be/YcK2vUpNt0g
Catalog No. 50-196-0298 Supplier Advanced Biomatrix Supplier No. 52251KIT
TeloCol-6 bovine collagen solution is derived from an acid extraction process yielding a telopeptide-intact collagen. The pro-peptide regions at both ends of the collagen chain, N- and C-telopeptide regions, are maintained. TeloCol-6 collagen is approximately 95% Type I collagen with the remainder being comprised of Type III collagen. Each product includes a bottle containing 50 ml of collagen solution accompanied with a bottle of pre-formulated neutralizing solution for the formation of a collagen gel. This product is supplied at a concentration of approximately 6 mg/ml with the concentration for each specific lot provided on the product label and Certificate of Analysis that is available with the purchase of each product. TeloCol-6 is sterile filtered and provided in user-friendly packaging for use and storage. TeloCol-6 is ideal for coating of surfaces, providing preparation of thin layers for culturing cells, or use as a solid gel. Collagen Video: https://youtu.be/YcK2vUpNt0g