Learn More
Advanced Biomatrix TeloCol-3, Solution, Acid Soluble, 3 mg/ml with Neutralization Solution
Supplier:  Advanced Biomatrix 50261KIT
Encompass_Preferred
TeloCol-3 is 50 mL of 3 mg/ml, type I bovine telocollagen solution for 2D coatings or 3D hydrogels. The kit comes with a neutralization solution for easy handling. Being telocollagen, TeloCol-3 will form firmer hydrogels, and gel significantly faster than atelocollagen solutions with the same concentration. TeloCol-3 bovine collagen solution is derived from an acid extraction process yielding a telopeptide-intact collagen. TeloCol is soluble telocollagen in 0.01 N HCL. The telopeptide regions at both ends of the collagen chain, N- and C-telopeptide regions, are maintained. TeloCol-3 collagen is approximately 95% Type I collagen with the remainder being comprised of Type III collagen. TeloCol-3 is sterile filtered. TeloCol-3 is ideal for coating of surfaces, providing preparation of thin layers for culturing cells, or use as a solid gel. Collagen Training Video: https://youtu.be/YcK2vUpNt0g
Catalog No. NC1382870