Learn More
Medchemexpress LLC HY-126482 5mg Medchemexpress, Telomerase-IN-2 CAS:1610878-54-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1264825MG
Encompass_Preferred
Medchemexpress, HY-126482 5mg Telomerase-IN-2 CAS:1610878-54-0 Telomerase-IN-2 is a telomerase inhibitor, and inhibits telomerase activity by decreasing expression of dyskerin, with an IC50 of 0.89 µM. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9731