Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chiron Telomeric Acid, FTCAs Standards
SDP

Catalog No. 14847850ME Shop All Spex CertiPrep Products
Change view
Click to view available options
Chemical Name or Material:
2H,2H,3H,3H-Perfluorodecanoic acid
2H,2H,3H,3H-Perfluorohexanoic acid
2H,2H,3H,3H-Perfluorooctanoic acid
2H,2H-Perfluorodecanoic acid
2H,2H-Perfluorooctanoic acid
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Chemical Name or Material
14-847-850ME 2H,2H,3H,3H-Perfluorooctanoic acid
85-481-050ME 2H,2H-Perfluorodecanoic acid
15-034-850ME 2H,2H-Perfluorooctanoic acid
15-417-1050ME 2H,2H,3H,3H-Perfluorodecanoic acid
28-306-50ME 2H,2H,3H,3H-Perfluorohexanoic acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-847-850ME Supplier Chiron Supplier No. 14847.850ME

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material 2H,2H,3H,3H-Perfluorooctanoic acid
CAS 914637-49-3
Concentration 50 μg/mL
Synonym 5:3 FTCA
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.