Learn More
Cayman Chemical TelomycIn 2.5mg
SDP
Catalog No. 502939823
Encompass_Preferred
A macrocyclic peptide lactone antibiotic; active against Gram-positive bacteria, including S. aureus, MRSA, and E. faecium (MICs = 4, 2, and 16 UG/ml, respectively); eradicates B. anserina spirochetosis infection in chicks
Catalog No. 50-293-9823 Supplier Cayman Chemical Supplier No. 281372.5 MG
A macrocyclic peptide lactone antibiotic; active against Gram-positive bacteria, including S. aureus, MRSA, and E. faecium (MICs = 4, 2, and 16 UG/ml, respectively); eradicates B. anserina spirochetosis infection in chicks