Learn More
Cayman Chemical Telotristat, Purity: ≥98%, CAS: 1033805-28-5, 10 mg
SDP
Catalog No. 502990167
Encompass_Preferred
A TPH1 inhibitor (IC50 = 16 nM); dietary administration to pregnant and lactating mice decreases 5-HT levels in maternal serum and decreases the femoral bone volume to tissue volume ratio, trabecular number and thickness, and cortical tissue mineral density, as well as serum calcium levels, in pups
Catalog No. 50-299-0167 Supplier Cayman Chemical Supplier No. 2942410 MG
A TPH1 inhibitor (IC50 = 16 nM); dietary administration to pregnant and lactating mice decreases 5-HT levels in maternal serum and decreases the femoral bone volume to tissue volume ratio, trabecular number and thickness, and cortical tissue mineral density, as well as serum calcium levels, in pups