Learn More
Medchemexpress LLC Telotristat ethyl | 1033805-22-9 | 98.9% | 1 ML
SDP
Catalog No. 5000338154
Encompass_Preferred
Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.
  • Reduces serotonin content in the periphery
  • Prevents increase in blood neutrophil counts
  • Provides significant protection in a mouse model of inflammatory bowel disease
  • Protects mouse IBD model confirmed by histopathology evaluation
  • Does not deplete enteric neuronal serotonin (5-HT) or alter constitutive gastrointestinal motility in mice
  • Alleviates severity of trinitrobenzene sulfonic acid (TNBS)-induced colitis
Catalog No. 50-003-38154 Supplier Medchemexpress LLC Supplier No. HY13055A10MM/1ML
Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.
  • Reduces serotonin content in the periphery
  • Prevents increase in blood neutrophil counts
  • Provides significant protection in a mouse model of inflammatory bowel disease
  • Protects mouse IBD model confirmed by histopathology evaluation
  • Does not deplete enteric neuronal serotonin (5-HT) or alter constitutive gastrointestinal motility in mice
  • Alleviates severity of trinitrobenzene sulfonic acid (TNBS)-induced colitis