Learn More
TARGETMOL CHEMICALS INC TELOTRISTAT ETIPRATE 25MG
SDP
Catalog No. 502579619
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telotristat Etiprate (LX 1606 Hippurate) is an orally bioavailable tryptophan hydroxylase (TPH) inhibitor with potential antiserotonergic activity. purity: 99%
Catalog No. 50-257-9619 Supplier TARGETMOL CHEMICALS INC Supplier No. T604425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Telotristat Etiprate (LX 1606 Hippurate) is an orally bioavailable tryptophan hydroxylase (TPH) inhibitor with potential antiserotonergic activity. purity: 99%