Learn More
Cayman Chemical TelratolImod 10mg
SDP
Catalog No. 502948413
Encompass_Preferred
A TLR7 and TLR8 agonist; increases the HA-induced secretion of IFN-γ in mouse splenocytes; enhances neutralizing antibody responses to a HIV-1 pseudovirus in rhesus macaques for at least one year; reduces tumor growth and increases metastasis-free survival in E0771 and 4T1.2 murine mammary carcinoma models at 0.8 mg/ml, intratumorally; increases intratumoral production of cytokines in a 4T1.2 murine model when administered intratumorally
Catalog No. 50-294-8413 Supplier Cayman Chemical Supplier No. 3317210 MG
A TLR7 and TLR8 agonist; increases the HA-induced secretion of IFN-γ in mouse splenocytes; enhances neutralizing antibody responses to a HIV-1 pseudovirus in rhesus macaques for at least one year; reduces tumor growth and increases metastasis-free survival in E0771 and 4T1.2 murine mammary carcinoma models at 0.8 mg/ml, intratumorally; increases intratumoral production of cytokines in a 4T1.2 murine model when administered intratumorally