Learn More
Medchemexpress LLC HY-109104 10mg Medchemexpress, Telratolimod CAS:1359993-59-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10910410MG
Encompass_Preferred
Medchemexpress, HY-109104 10mg Telratolimod CAS:1359993-59-1 Telratolimod is a toll like receptors 7/8 (TLR7/8) agonist, with antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8681