Learn More
Selleck Chemical LLC Telratolimod 5mg
SDP
Supplier:  Selleck Chemical LLC S96235MG
Encompass_Preferred
Telratolimod (3M-052, MEDI9197) is an agonist of toll-like receptor 7 (TLR-7) and toll-like receptor 8 (TLR-8) with antitumor activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3480