Learn More
Electron Microscopy Sciences Omniprobe TEM, Grid Dual Holder, Aluminum
SDP
Catalog No. 5033772
Encompass
Post Base TEM Grid Holder. Short post – standard is 1/8" (3.2 mm) diameter x 0.15" (4 mm) length.  Aluminum is non-magnetic.
Catalog No. 50-337-72 Supplier Electron Microscopy Sciences Supplier No. 75968AL
Post Base TEM Grid Holder. Short post – standard is 1/8" (3.2 mm) diameter x 0.15" (4 mm) length.  Aluminum is non-magnetic.