Learn More
Electron Microscopy Sciences Omniprobe TEM, Grid Dual Holder, Stainless steel
SDP
Catalog No. 5033773
Encompass
Post Base TEM Grid Holder. Short post – standard is 1/8" (3.2 mm) diameter x 0.15" (4 mm) length.  Stainless Steel is very slightly magnetic.
Catalog No. 50-337-73 Supplier Electron Microscopy Sciences Supplier No. 75968SS
Post Base TEM Grid Holder. Short post – standard is 1/8" (3.2 mm) diameter x 0.15" (4 mm) length.  Stainless Steel is very slightly magnetic.