Learn More
Electron Microscopy Sciences Omniprobe TEM Grid & Sample Holder, Stainless steel
SDP
Catalog No. 5033771
Encompass
This station is meant for 2 TEM grids and 1 sample Pin Stub. Long post – standard is 1/8" (3.2 mm) diameter x 0.32" (8.1 mm) length. Comes with an aluminum sample stub. Stainless Steel is very slightly magnetic.
Catalog No. 50-337-71 Supplier Electron Microscopy Sciences Supplier No. 75971SS
This station is meant for 2 TEM grids and 1 sample Pin Stub. Long post – standard is 1/8" (3.2 mm) diameter x 0.32" (8.1 mm) length. Comes with an aluminum sample stub. Stainless Steel is very slightly magnetic.