Learn More
Simpore Inc 400 nm thick Non-porous Silicon NitrideSingle 100x500 µm window with free edge200 nm thick gold contacts2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818925
Encompass_Preferred
400 nm thick Non-porous Silicon NitrideSingle 100x500 µm window with free edge200 nm thick gold contacts2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818925 Supplier Simpore Inc Supplier No. SN100LFTAU
400 nm thick Non-porous Silicon NitrideSingle 100x500 µm window with free edge200 nm thick gold contacts2.9 mm hexagonal + 0.1 mm thick chip framePack of 10