Learn More
Simpore Inc 400 nm thick Non-porous Silicon NitrideSingle 100x500 µm window with free edge2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818921
Encompass_Preferred
400 nm thick Non-porous Silicon NitrideSingle 100x500 µm window with free edge2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818921 Supplier Simpore Inc Supplier No. SN100LFT
400 nm thick Non-porous Silicon NitrideSingle 100x500 µm window with free edge2.9 mm hexagonal + 0.1 mm thick chip framePack of 10