Learn More
NANOPROBES INC NanoVan, 5 mL
Catalog No. NC0659245
Encompass
A 2% Vanadium negative stain recommended for use with Nanogold or other applications where a low atomic number light stain is needed.
Catalog No. NC0659245 Supplier NANOPROBES INC Supplier No. 20115ML
A 2% Vanadium negative stain recommended for use with Nanogold or other applications where a low atomic number light stain is needed.