Learn More
NANOPROBES INC Nano-W, 5 mL
Catalog No. NC0659246
Encompass
A 2% negative stain based on an organotungsten compound. High density, high contrast.
Catalog No. NC0659246 Supplier NANOPROBES INC Supplier No. 20185ML
A 2% negative stain based on an organotungsten compound. High density, high contrast.