Learn More
Simpore Inc 5 nm thick Non-porous Amorphous Silicon25×25 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819079
Encompass_Preferred
5 nm thick Non-porous Amorphous Silicon25×25 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819079 Supplier Simpore Inc Supplier No. US100A05Q00
5 nm thick Non-porous Amorphous Silicon25×25 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10