Learn More
Simpore Inc 25 nm thick Non-porous <1-0-0> Crystal Silicon(8) 50x50 µM + (1) 50x100 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819081
Encompass_Preferred
25 nm thick Non-porous <1-0-0> Crystal Silicon(8) 50x50 µM + (1) 50x100 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819081 Supplier Simpore Inc Supplier No. US100C25Q33A
25 nm thick Non-porous <1-0-0> Crystal Silicon(8) 50x50 µM + (1) 50x100 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10