Learn More
Simpore Inc 9 nm thick Non-porous Amorphous Silicon(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819082
Encompass_Preferred
9 nm thick Non-porous Amorphous Silicon(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819082 Supplier Simpore Inc Supplier No. US100HA09Q33
9 nm thick Non-porous Amorphous Silicon(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10