Learn More
Simpore Inc 5 nm thick Non-porous Amorphous Silicon(8) 50x50 µM + (1) 50x100 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819080
Encompass_Preferred
5 nm thick Non-porous Amorphous Silicon(8) 50x50 µM + (1) 50x100 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819080 Supplier Simpore Inc Supplier No. US100A05Q33A
5 nm thick Non-porous Amorphous Silicon(8) 50x50 µM + (1) 50x100 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10