Learn More
Simpore Inc 15 nm thick Non-porous Amorphous Silicon(2) 100x1,500 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819072
Encompass_Preferred
15 nm thick Non-porous Amorphous Silicon(2) 100x1,500 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2819072 Supplier Simpore Inc Supplier No. US100A15L
15 nm thick Non-porous Amorphous Silicon(2) 100x1,500 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10