Learn More
Electron Microscopy Sciences TEM Screen Recoating Hitachi H7000, H8000 Outer
SDP
Catalog No. 5029984
Encompass
For optimum viewing, fluorescent screens should be recoated regularly. Viewing screens are easily damaged by the electron beam, especially at high accelerating voltages where knock-on damage can be severe. A proprietary process where 1 to 2 micron particles of phosphor are deposited on the screen and produce the highest possible resolution.
Catalog No. 50-299-84 Supplier Electron Microscopy Sciences Supplier No. 2510
For optimum viewing, fluorescent screens should be recoated regularly. Viewing screens are easily damaged by the electron beam, especially at high accelerating voltages where knock-on damage can be severe. A proprietary process where 1 to 2 micron particles of phosphor are deposited on the screen and produce the highest possible resolution.