Learn More
Electron Microscopy Sciences TEM Screen Recoating Hitachi Focusing
SDP
Catalog No. 5030010
Encompass
For optimum viewing, fluorescent screens should be recoated regularly. Viewing screens are easily damaged by the electron beam, especially at high accelerating voltages where knock-on damage can be severe. A proprietary process where 1 to 2 micron particles of phosphor are deposited on the screen and produce the highest possible resolution.
Catalog No. 50-300-10 Supplier Electron Microscopy Sciences Supplier No. 2520
For optimum viewing, fluorescent screens should be recoated regularly. Viewing screens are easily damaged by the electron beam, especially at high accelerating voltages where knock-on damage can be severe. A proprietary process where 1 to 2 micron particles of phosphor are deposited on the screen and produce the highest possible resolution.