Learn More
Simpore Inc 40 nm thick Non-porous Silicon Dioxide(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819068
Encompass_Preferred
40 nm thick Non-porous Silicon Dioxide(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819068 Supplier Simpore Inc Supplier No. SO100HA40Q33
40 nm thick Non-porous Silicon Dioxide(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10