Learn More
Simpore Inc 20 nm thick Non-porous Silicon Dioxide(8) 50x50 µm + (1) 50x100 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819067
Encompass_Preferred
20 nm thick Non-porous Silicon Dioxide(8) 50x50 µm + (1) 50x100 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819067 Supplier Simpore Inc Supplier No. SO100HA20Q33A
20 nm thick Non-porous Silicon Dioxide(8) 50x50 µm + (1) 50x100 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10