Learn More
Simpore Inc 50 nm thick Non-porous Silicon Nitride500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818909
Encompass_Preferred
50 nm thick Non-porous Silicon Nitride500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818909 Supplier Simpore Inc Supplier No. SN100A50Q05
50 nm thick Non-porous Silicon Nitride500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10