Learn More
Simpore Inc 5 nm thick Non-porous Silicon Nitride(2) 50x1,500 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818889
Encompass_Preferred
5 nm thick Non-porous Silicon Nitride(2) 50x1,500 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818889 Supplier Simpore Inc Supplier No. SN100A05L
5 nm thick Non-porous Silicon Nitride(2) 50x1,500 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10