Learn More
Simpore Inc 5 nm thick Non-porous Silicon Nitride25×25 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818895
Encompass_Preferred
5 nm thick Non-porous Silicon Nitride25×25 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818895 Supplier Simpore Inc Supplier No. SN100A05Q00
5 nm thick Non-porous Silicon Nitride25×25 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10