Learn More
Simpore Inc 20 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818897
Encompass_Preferred
20 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818897 Supplier Simpore Inc Supplier No. SN100A20MP2Q05
20 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10