Learn More
Simpore Inc 50 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818899
Encompass_Preferred
50 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818899 Supplier Simpore Inc Supplier No. SN100A50MP2Q05
50 nm thick Microporous Silicon Nitride2 µm diameter pores500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10