Learn More
Simpore Inc 50 nm thick Non-porous Silicon Nitride100×100 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818911
Encompass_Preferred
50 nm thick Non-porous Silicon Nitride100×100 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818911 Supplier Simpore Inc Supplier No. SN100A50Q01
50 nm thick Non-porous Silicon Nitride100×100 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10