Learn More
Simpore Inc 50 nm thick Non-porous Silicon Nitride1,000×1,000 µm single window2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818917
Encompass_Preferred
50 nm thick Non-porous Silicon Nitride1,000×1,000 µm single window2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818917 Supplier Simpore Inc Supplier No. SN100HA50Q10
50 nm thick Non-porous Silicon Nitride1,000×1,000 µm single window2.9 mm hexagonal + 0.1 mm thick chip framePack of 10