Learn More
Simpore Inc 20 nm thick Nanoporous Silicon Nitride15-20 nm diameter pores(8) 100x100 µm + (1) 100x350 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818930
Encompass_Preferred
20 nm thick Nanoporous Silicon Nitride15-20 nm diameter pores(8) 100x100 µm + (1) 100x350 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818930 Supplier Simpore Inc Supplier No. SN100P20Q33PK
20 nm thick Nanoporous Silicon Nitride15-20 nm diameter pores(8) 100x100 µm + (1) 100x350 µm windows2.9 mm diameter + 0.1 mm thick chip framePack of 10