Learn More
Simpore Inc 5 nm thick Non-porous Silicon Nitride(8) 50x50 µm + (1) 50x100 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818933
Encompass_Preferred
5 nm thick Non-porous Silicon Nitride(8) 50x50 µm + (1) 50x100 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2818933 Supplier Simpore Inc Supplier No. SN100HA05Q33APK
5 nm thick Non-porous Silicon Nitride(8) 50x50 µm + (1) 50x100 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10