Learn More
Simpore Inc 10 nm thick Non-porous Silicon Nitride(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819055
Encompass_Preferred
10 nm thick Non-porous Silicon Nitride(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819055 Supplier Simpore Inc Supplier No. SN100HA10Q33
10 nm thick Non-porous Silicon Nitride(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10