Learn More
Simpore Inc 10 nm thick Non-porous Silicon Nitride(8) 250x250 µm + (1) 250x500 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819061
Encompass_Preferred
10 nm thick Non-porous Silicon Nitride(8) 250x250 µm + (1) 250x500 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819061 Supplier Simpore Inc Supplier No. SN100HA10Q33B
10 nm thick Non-porous Silicon Nitride(8) 250x250 µm + (1) 250x500 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10