Learn More
Simpore Inc 20 nm thick Non-porous Silicon Nitride(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819064
Encompass_Preferred
20 nm thick Non-porous Silicon Nitride(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10
Catalog No. NC2819064 Supplier Simpore Inc Supplier No. SN100HA20Q33
20 nm thick Non-porous Silicon Nitride(8) 100x100 µm + (1) 100x350 µm windows2.9 mm hexagonal + 0.1 mm thick chip framePack of 10