Learn More
Simpore Inc 20 nm thick Non-porous Silicon Nitride500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818903
Encompass_Preferred
20 nm thick Non-porous Silicon Nitride500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10
Catalog No. NC2818903 Supplier Simpore Inc Supplier No. SN100HA20Q05
20 nm thick Non-porous Silicon Nitride500×500 µm single window2.9 mm diameter + 0.1 mm thick chip framePack of 10