Learn More
Electron Microscopy Sciences TEM Specimen Supports Molybdenum 200 mesh
SDP
Catalog No. 5031073
Encompass_Preferred
We have extended our range of TEM grid materials to include four types, which are now available in Molybdenum. The new products are manufactured using a process known as chemical ʽmilling' (etching) instead of the more familiar technique of ʽelectroforming' (deposition) that is used in the manufacture of copper, nickel and gold products. Molybdenum is used principally in applications where it's high temperature, hardness, expansion of coefficient and corrosion resistance characteristics are considered important. The material which is used has a purity of 99.9%.
Pitch: 125 µm
Hole: 90 µm
Bar: 35 µm
Thickness: 25 µm
Catalog No. 50-310-73 Supplier Electron Microscopy Sciences Supplier No. G200MO
We have extended our range of TEM grid materials to include four types, which are now available in Molybdenum. The new products are manufactured using a process known as chemical ʽmilling' (etching) instead of the more familiar technique of ʽelectroforming' (deposition) that is used in the manufacture of copper, nickel and gold products. Molybdenum is used principally in applications where it's high temperature, hardness, expansion of coefficient and corrosion resistance characteristics are considered important. The material which is used has a purity of 99.9%.
Pitch: 125 µm
Hole: 90 µm
Bar: 35 µm
Thickness: 25 µm