Learn More
Electron Microscopy Sciences TEM Specimen Supports Molybdenum 1000 µm
SDP
Catalog No. 5031074
Encompass_Preferred
We have extended our range of TEM grid materials to include four types, which are now available in Molybdenum. The new products are manufactured using a process known as chemical ʽmilling' (etching) instead of the more familiar technique of ʽelectroforming' (deposition) that is used in the manufacture of copper, nickel and gold products. Molybdenum is used principally in applications where it's high temperature, hardness, expansion of coefficient and corrosion resistance characteristics are considered important. The material which is used has a purity of 99.9%.
Hole: 1000 µm
Thickness: 50 µm
Catalog No. 50-310-74 Supplier Electron Microscopy Sciences Supplier No. G1000MO
We have extended our range of TEM grid materials to include four types, which are now available in Molybdenum. The new products are manufactured using a process known as chemical ʽmilling' (etching) instead of the more familiar technique of ʽelectroforming' (deposition) that is used in the manufacture of copper, nickel and gold products. Molybdenum is used principally in applications where it's high temperature, hardness, expansion of coefficient and corrosion resistance characteristics are considered important. The material which is used has a purity of 99.9%.
Hole: 1000 µm
Thickness: 50 µm