Learn More
Medchemexpress LLC Temafloxacin | 108319-06-8 | 99.65% | 417.38 | 5 MG
SDP
Catalog No. 5000390427
Encompass_Preferred
Temafloxacin (TMFX) is an orally active quinolone broad-spectrum antibacterial agent. It is well tolerated in lower respiratory and genitourinary tract infections, demonstrating good antibacterial activity against gram-positive and gram-negative bacteria, with specific MIC ranges for E. coli, P. aeruginosa, and S. aureus. It shows good inhibitory activity in murine pyelonephritis and, as the hydrochloride form, rapid gastrointestinal absorption with excellent tissue and body fluid penetration.
  • Orally active quinolone broad-spectrum antibacterial agent
  • Well tolerated in lower respiratory and genitourinary tract infections
  • Exhibits good antibacterial activity against gram-positive and gram-negative bacteria
  • Demonstrates good inhibitory activity against murine pyelonephritis
  • Rapid gastrointestinal absorption and excellent tissue and body fluid penetration
Catalog No. 50-003-90427 Supplier Medchemexpress LLC Supplier No. HY164875MG
Temafloxacin (TMFX) is an orally active quinolone broad-spectrum antibacterial agent. It is well tolerated in lower respiratory and genitourinary tract infections, demonstrating good antibacterial activity against gram-positive and gram-negative bacteria, with specific MIC ranges for E. coli, P. aeruginosa, and S. aureus. It shows good inhibitory activity in murine pyelonephritis and, as the hydrochloride form, rapid gastrointestinal absorption with excellent tissue and body fluid penetration.
  • Orally active quinolone broad-spectrum antibacterial agent
  • Well tolerated in lower respiratory and genitourinary tract infections
  • Exhibits good antibacterial activity against gram-positive and gram-negative bacteria
  • Demonstrates good inhibitory activity against murine pyelonephritis
  • Rapid gastrointestinal absorption and excellent tissue and body fluid penetration