Learn More
Cayman Chemical Temafloxacin, Purity: ≥95%, CAS: 108319-06-8, 10 mg
SDP
Catalog No. 502979697
Encompass_Preferred
A fluoroquinolone antibiotic that inhibits DNA topoisomerase inhibitor; broad antimicrobial activity against gram-positive and gram-negative bacteria, such as S. pneumoniae, M. hominis, and anaerobic bacteria, including B. fragilis
Catalog No. 50-297-9697 Supplier Cayman Chemical Supplier No. 1735210 MG
A fluoroquinolone antibiotic that inhibits DNA topoisomerase inhibitor; broad antimicrobial activity against gram-positive and gram-negative bacteria, such as S. pneumoniae, M. hominis, and anaerobic bacteria, including B. fragilis